Assay Details
| Gene Symbol: Uqcrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025899.2 | NP_080175.1 | 10 - 11 | 1061 | 74 | |
| GenBank mRNA | AK004492.1 | - | 10 - 11 | 1037 | 74 | |
| AK005151.1 | - | 10 - 11 | 1045 | 74 | ||
| AK007669.1 | - | 9 - 10 | 925 | 74 | ||
| AK075856.1 | - | 10 - 11 | 1005 | 74 | ||
| AK088103.1 | - | 10 - 11 | 1015 | 74 | ||
| BC003423.1 | - | 10 - 11 | 1033 | 2609295 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uqcrc2
Genomic Map