Assay Details
| Gene Symbol: Uqcrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025899.2 | NP_080175.1 | 11 - 12 | 1146 | 92 | |
| GenBank mRNA | AK003975.1 | - | 11 - 12 | 1116 | 92 | |
| AK004492.1 | - | 11 - 12 | 1122 | 92 | ||
| AK005151.1 | - | 11 - 12 | 1130 | 92 | ||
| AK007669.1 | - | 10 - 11 | 1010 | 92 | ||
| AK075856.1 | - | 11 - 12 | 1090 | 92 | ||
| AK077583.1 | - | 11 - 12 | 1095 | 92 | ||
| AK088103.1 | - | 11 - 12 | 1100 | 92 | ||
| AK167377.1 | - | 11 - 12 | 1099 | 92 | ||
| BC003423.1 | - | 11 - 12 | 1118 | 2609295 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uqcrc2
Genomic Map