Assay Details
| Gene Symbol: Nptxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF316612.1 | - | 1 - 2 | 719 | 118 | |
| AY394487.1 | - | 2 - 3 | 445 | 118 | ||
| BC141096.1 | - | 1 - 2 | 716 | 9055708 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nptxr
Genomic Map