Assay Details
| Gene Symbol: Uck2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030724.3 | NP_109649.1 | 5 - 6 | 847 | 62 | |
| GenBank mRNA | AF236636.1 | - | 5 - 6 | 852 | 62 | |
| AK051894.1 | - | 5 - 6 | 617 | 62 | ||
| AK053745.1 | - | 5 - 6 | 807 | 62 | ||
| AK082837.1 | - | 5 - 6 | 810 | 62 | ||
| AK155939.1 | - | 5 - 6 | 811 | 62 | ||
| AK168318.1 | - | 5 - 6 | 825 | 62 | ||
| BC023789.1 | - | 5 - 6 | 811 | 5352192 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uck2
Genomic Map