Assay Details
| Gene Symbol: Clic1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033444.2 | NP_254279.1 | 4 - 5 | 81 | ||
| GenBank mRNA | AK088809.1 | - | 4 - 5 | 81 | ||
| AK150382.1 | - | 4 - 5 | 81 | |||
| AK150611.1 | - | 4 - 5 | 81 | |||
| AK153509.1 | - | 4 - 5 | 81 | |||
| AK167765.1 | - | 4 - 5 | 81 | |||
| AK168262.1 | - | 4 - 5 | 403 | 81 | ||
| BC004658.1 | - | 4 - 5 | 3498118 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Clic1
Genomic Map