Assay Details
| Gene Symbol: Cox4i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053091.2 | NP_444321.1 | 1 - 2 | 58 | 84 | |
| GenBank mRNA | AF271382.1 | - | 1 - 2 | 58 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AF271382.1, NM_053091.2 transcripts.
Set Membership: |
|

Cox4i2
Genomic Map