Assay Details
| Gene Symbol: Cox4i2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053091.2 | NP_444321.1 | 4 - 5 | 432 | 84 | |
| GenBank mRNA | AF271382.1 | - | 4 - 5 | 432 | 84 | |
| BC049623.1 | - | 3 - 4 | 441 | 6532529 | 84 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054663 | NM_053091.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054663 | AF271382.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2054663 | BC049623.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 858992 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 230809 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 54176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192560 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970459 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 220199 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 64259 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171481 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123307 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054663 | NM_053091.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054663 | AF271382.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2054663 | BC049623.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P202942 | NM_053091.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P202942 | AF271382.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750977 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750973 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750984 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21750972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21751010 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04742668 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123307 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171481 | - | - |
| Whole Mouse Genome G4122A | A_51_P202942 | NM_053091.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P202942 | AF271382.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128625 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119395 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171481 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123307 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02198704 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01676358 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01231662 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01676350 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01676357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751000 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751085 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751009 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751069 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21750987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21751021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01552123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02262401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01552126 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750972 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01406064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02212722 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01636045 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01422103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01636056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01629824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751004 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01231662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01604252 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01676358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01636049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01948528 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01636055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751029 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01676354 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01676357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01960286 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01375083 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751000 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750987 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01176028 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21751006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21750994 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10477185 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477184 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477183 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477182 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477181 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477186 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10477180 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1421373_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10477179 | - | - |
| Mouse Genome 430 2.0 Array | 1421373_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1421373_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 84682_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 84682_PM_at | - | - |
| Mouse Expression Set 430 | 1421373_PM_at | - | - |
| Mouse Expression Set 430 | 1421373_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421373_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1421373_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421373_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1421373_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 551351 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6370519 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006370519 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2612178 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2612178 | - | - |
| mouse-6 v1.1 (Illumina) | scl20106.5.1_101-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6370519 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl20106.5.1_101-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6370519 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2612178 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2612178 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl20106.5.1_101-S | - | - |

Cox4i2
Genomic Map