Assay Details
| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293313.1 | NP_001280242.1 | 6 - 7 | 744 | 119 | |
| NM_138314.4 | NP_612187.2 | 6 - 7 | 747 | 119 | ||
| NM_178071.6 | NP_835172.1 | 6 - 7 | 744 | 119 | ||
| GenBank mRNA | AF202048.1 | - | 5 - 6 | 648 | 119 | |
| AK044962.1 | - | 6 - 7 | 718 | 119 | ||
| AK085222.1 | - | 6 - 7 | 692 | 119 | ||
| AK133221.1 | - | 1215 | 119 | |||
| AK144915.1 | - | 6 - 7 | 748 | 119 | ||
| AK164652.1 | - | 6 - 7 | 692 | 119 | ||
More Information
Set Membership: |
|

Nme7
Genomic Map