Assay Details
| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293313.1 | NP_001280242.1 | 11 - 12 | 1243 | 122 | |
| NM_138314.4 | NP_612187.2 | 11 - 12 | 1246 | 122 | ||
| GenBank mRNA | AF202048.1 | - | 10 - 11 | 1147 | 122 | |
| AK133221.1 | - | 1714 | 122 | |||
| AK144915.1 | - | 11 - 12 | 1247 | 122 | ||
| AK160566.1 | - | 3 - 4 | 202 | 122 | ||
| BC038021.1 | - | 12 - 13 | 1273 | 3963844 | 122 | |
| BU609451.1 | - | 3 - 4 | 122 | |||
| CD775820.1 | - | 3 - 4 | 122 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nme7
Genomic Map