Assay Details
| Gene Symbol: Gusb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010368.1 | NP_034498.1 | 1 - 2 | 220 | 76 | |
| GenBank mRNA | AK002832.1 | - | 1 - 2 | 266 | 76 | |
| AK041058.1 | - | 1 - 2 | 221 | 76 | ||
| AK076109.1 | - | 1 - 2 | 252 | 76 | ||
| AK136519.1 | - | 1 - 2 | 277 | 76 | ||
| AK146247.1 | - | 1 - 2 | 442 | 76 | ||
| AK149814.1 | - | 1 - 2 | 250 | 76 | ||
| AK150048.1 | - | 1 - 2 | 221 | 76 | ||
| AK151802.1 | - | 1 - 2 | 221 | 76 | ||
| AK152206.1 | - | 1 - 2 | 221 | 76 | ||
| AK159163.1 | - | 1 - 2 | 221 | 76 | ||
| AK159526.1 | - | 1 - 2 | 221 | 76 | ||
| AK159564.1 | - | 1 - 2 | 212 | 76 | ||
| AK159804.1 | - | 1 - 2 | 221 | 76 | ||
| AK162436.1 | - | 1 - 2 | 221 | 76 | ||
| BC004616.1 | - | 1 - 2 | 257 | 76 | ||
| BC071226.1 | - | 1 - 2 | 241 | 5325429 | 76 | |
| J03047.1 | - | 1 - 2 | 220 | 76 | ||
| M19279.1 | - | 1 - 2 | 220 | 76 | ||
| M28540.1 | - | 1 - 2 | 220 | 76 | ||
| M28541.1 | - | 1 - 2 | 220 | 76 | ||
| M63836.1 | - | 1 - 2 | 220 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Citations (15)
Homeodomain transcription factor NKX2.2 functions in immature cells to control enteroendocrine differentiation and is expressed in gastrointestinal neuroendocrine tumors.
- Authors
- Wang YC, Gallego-Arteche E, Iezza G, Yuan X, Matli MR, Choo SP, Zuraek MB, Gogia R, Lynn FC, German MS, Bergsland EK, Donner DB, Warren RS, Nakakura EK
- Journal
- Endocr Relat Cancer - Page No 267 (Sun Mar 01 00:00:00 UTC 2009)
- Volume (Year)
- 16 (2009)
- Product Usage
- -
Blockade of IP-10/CXCR3 promotes progressive renal fibrosis.
- Authors
- Nakaya I, Wada T, Furuichi K, Sakai N, Kitagawa K, Yokoyama H, Ishida Y, Kondo T, Sugaya T, Kawachi H, Shimizu F, Narumi S, Haino M, Gerard C, Matsushima K, Kaneko S
- Journal
- Nephron Exp Nephrol - Page No e12 (Mon Jan 01 00:00:00 UTC 2007)
- Volume (Year)
- 107 (2007)
- Product Usage
- -
INrf2 (Keap1) targets Bcl-2 degradation and controls cellular apoptosis.
- Authors
- Niture SK, Jaiswal AK
- Journal
- Cell Death Differ - Page No (Fri Sep 24 00:00:00 UTC 2010)
- Volume (Year)
- (2010)
- Product Usage
- -
Tamoxifen administration routes and dosage for inducible Cre-mediated gene disruption in mouse hearts.
- Authors
- Andersson KB, Winer LH, Mork HK, Molkentin JD, Jaisser F
- Journal
- Transgenic Res - Page No 715 (Sun Aug 01 00:00:00 UTC 2010)
- Volume (Year)
- 19 (2010)
- Product Usage
- -
Knockdown of the Dnmt1s transcript using small interfering RNA in primary murine and bovine fibroblast cells.
- Authors
- Adams AM, Pratt SL, Stice SL
- Journal
- Mol Reprod Dev - Page No 311 (Tue Nov 01 00:00:00 UTC 2005)
- Volume (Year)
- 72 (2005)
- Product Usage
- -
Dendritic cells contribute to autoimmune kidney injury in MRL-Faslpr mice.
- Authors
- Iwata Y, Furuichi K, Sakai N, Yamauchi H, Shinozaki Y, Zhou H, Kurokawa Y, Toyama T, Kitajima S, Okumura T, Yamada S, Maruyama I, Matsushima K, Kaneko S, Wada T
- Journal
- J Rheumatol - Page No 306 (Sun Feb 01 00:00:00 UTC 2009)
- Volume (Year)
- 36 (2009)
- Product Usage
- -
Selection of reference genes for quantitative real-time RT-PCR studies in mouse brain.
- Authors
- Boda E, Pini A, Hoxha E, Parolisi R, Tempia F
- Journal
- J Mol Neurosci - Page No 238 (Sun Mar 01 00:00:00 UTC 2009)
- Volume (Year)
- 37 (2009)
- Product Usage
- -
Puma and Trail/Dr5 pathways control radiation-induced apoptosis in distinct populations of testicular progenitors.
- Authors
- Coureuil M, Ugolin N, Tavernier M, Chevillard S, Barroca V, Fouchet P, Allemand I
- Journal
- Plos One - Page No e12134 (Fri Jan 01 00:00:00 UTC 2010)
- Volume (Year)
- 5 (2010)
- Product Usage
- -
Protective effect of Juzen-taiho-to on hepatocarcinogenesis is mediated through the inhibition of Kupffer cell-induced oxidative stress.
- Authors
- Tsuchiya M, Kono H, Matsuda M, Fujii H, Rusyn I
- Journal
- Int J Cancer - Page No 2503 (Mon Dec 01 00:00:00 UTC 2008)
- Volume (Year)
- 123 (2008)
- Product Usage
- -
Molecular staging of epithelial maturation using secretory cell-specific genes as markers.
- Authors
- Zemke AC, Snyder JC, Brockway BL, Drake JA, Reynolds SD, Kaminski N, Stripp BR
- Journal
- Am J Respir Cell Mol Biol - Page No 340 (Sun Mar 01 00:00:00 UTC 2009)
- Volume (Year)
- 40 (2009)
- Product Usage
- -
Fumonisin B1-induced neurodegeneration in mice after intracerebroventricular infusion is concurrent with disruption of sphingolipid metabolism and activation of proinflammatory signaling.
- Authors
- Osuchowski MF, Edwards GL, Sharma RP
- Journal
- Neurotoxicology - Page No 211 (Tue Mar 01 00:00:00 UTC 2005)
- Volume (Year)
- 26 (2005)
- Product Usage
- -
Characterization of survival and phenotype throughout the life span in UCP2/UCP3 genetically altered mice.
- Authors
- McDonald RB, Walker KM, Warman DB, Griffey SM, Warden CH, Ramsey JJ, Horwitz BA
- Journal
- Exp Gerontol - Page No 1061 (Mon Dec 01 00:00:00 UTC 2008)
- Volume (Year)
- 43 (2008)
- Product Usage
- -
The ETS oncogene family transcription factor FEV identifies serotonin-producing cells in normal and neoplastic small intestine.
- Authors
- Wang YC, Zuraek MB, Kosaka Y, Ota Y, German MS, Deneris ES, Bergsland EK, Donner DB, Warren RS, Nakakura EK
- Journal
- Endocr Relat Cancer - Page No 283 (Mon Mar 01 00:00:00 UTC 2010)
- Volume (Year)
- 17 (2010)
- Product Usage
- -
An autoregulatory loop between Nrf2 and Cul3-Rbx1 controls their cellular abundance.
- Authors
- Kaspar JW, Jaiswal AK
- Journal
- J Biol Chem - Page No 21349 (Fri Jul 09 00:00:00 UTC 2010)
- Volume (Year)
- 285 (2010)
- Product Usage
- -
Evidence that SOX2 overexpression is oncogenic in the lung.
- Authors
- Lu Y, Futtner C, Rock JR, Xu X, Whitworth W, Hogan BL, Onaitis MW
- Journal
- Plos One - Page No e11022 (Fri Jan 01 00:00:00 UTC 2010)
- Volume (Year)
- 5 (2010)
- Product Usage
- -

Gusb
Genomic Map