Assay Details
| Gene Symbol: Gusb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010368.1 | NP_034498.1 | 2 - 3 | 409 | 91 | |
| GenBank mRNA | AK076109.1 | - | 2 - 3 | 441 | 91 | |
| AK136519.1 | - | 2 - 3 | 466 | 91 | ||
| AK146247.1 | - | 2 - 3 | 631 | 91 | ||
| AK149814.1 | - | 2 - 3 | 439 | 91 | ||
| AK150048.1 | - | 2 - 3 | 410 | 91 | ||
| AK151802.1 | - | 2 - 3 | 410 | 91 | ||
| AK152206.1 | - | 2 - 3 | 410 | 91 | ||
| AK159163.1 | - | 2 - 3 | 410 | 91 | ||
| AK159526.1 | - | 2 - 3 | 410 | 91 | ||
| AK159564.1 | - | 2 - 3 | 401 | 91 | ||
| AK159804.1 | - | 2 - 3 | 410 | 91 | ||
| AK162436.1 | - | 2 - 3 | 410 | 91 | ||
| BC004616.1 | - | 2 - 3 | 446 | 91 | ||
| BC018202.1 | - | 2 - 3 | 546 | 91 | ||
| BC071226.1 | - | 2 - 3 | 430 | 5325429 | 91 | |
| J03047.1 | - | 2 - 3 | 409 | 91 | ||
| M19279.1 | - | 2 - 3 | 409 | 91 | ||
| M28540.1 | - | 2 - 3 | 409 | 91 | ||
| M28541.1 | - | 2 - 3 | 409 | 91 | ||
| M63836.1 | - | 2 - 3 | 409 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gusb
Genomic Map