Assay Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007644.3 | NP_031670.1 | 4 - 5 | 908 | 62 | |
| GenBank mRNA | AB008553.1 | - | 4 - 5 | 813 | 62 | |
| AK011123.1 | - | 4 - 5 | 838 | 62 | ||
| AK083038.1 | - | 4 - 5 | 909 | 62 | ||
| AK144235.1 | - | 4 - 5 | 940 | 62 | ||
| BC029073.1 | - | 4 - 5 | 816 | 3962669 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scarb2
Genomic Map