Assay Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007644.3 | NP_031670.1 | 5 - 6 | 1008 | 113 | |
| GenBank mRNA | AB008553.1 | - | 5 - 6 | 913 | 113 | |
| AK011123.1 | - | 5 - 6 | 938 | 113 | ||
| AK083038.1 | - | 5 - 6 | 1009 | 113 | ||
| AK144235.1 | - | 5 - 6 | 1040 | 113 | ||
| AK149102.1 | - | 955 | 113 | |||
| BC029073.1 | - | 5 - 6 | 916 | 3962669 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scarb2
Genomic Map