Assay Details
| Gene Symbol: Scarb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007644.3 | NP_031670.1 | 9 - 10 | 1490 | 83 | |
| GenBank mRNA | AB008553.1 | - | 9 - 10 | 1395 | 83 | |
| AK011123.1 | - | 9 - 10 | 1420 | 83 | ||
| AK083038.1 | - | 9 - 10 | 1491 | 83 | ||
| AK144235.1 | - | 9 - 10 | 1522 | 83 | ||
| AK170555.1 | - | 150 | 83 | |||
| BC029073.1 | - | 9 - 10 | 1398 | 3962669 | 83 | |
More Information
Set Membership: |
|

Scarb2
Genomic Map