Assay Details
| Gene Symbol: Pola2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164057.1 | NP_001157529.1 | 14 - 15 | 1658 | 63 | |
| NM_008893.3 | NP_032919.2 | 14 - 15 | 1658 | 63 | ||
| GenBank mRNA | AK087989.1 | - | 14 - 15 | 1654 | 63 | |
| AK133832.1 | - | 14 - 15 | 1644 | 63 | ||
| AK150099.1 | - | 14 - 15 | 1600 | 63 | ||
| BC021424.1 | - | 14 - 15 | 1633 | 5011204 | 63 | |
| BC064795.1 | - | 14 - 15 | 1574 | 30111287 | 63 | |
| D13546.1 | - | 14 - 15 | 1591 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pola2
Genomic Map