Assay Details
| Gene Symbol: Ppl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008909.2 | NP_032935.2 | 3 - 4 | 460 | 73 | |
| GenBank mRNA | AF126834.1 | - | 3 - 4 | 400 | 73 | |
| AK142562.1 | - | 3 - 4 | 455 | 73 | ||
| BC057152.1 | - | 3 - 4 | 389 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppl
Genomic Map