Assay Details
| Gene Symbol: Ppox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008911.2 | NP_032937.1 | 9 - 10 | 1311 | 59 | |
| GenBank mRNA | AK134341.1 | - | 2743 | 59 | ||
| AK142632.1 | - | 9 - 10 | 1263 | 59 | ||
| BC002047.1 | - | 9 - 10 | 1266 | 3484079 | 59 | |
| D45185.1 | - | 9 - 10 | 1037 | 59 | ||
| U25114.1 | - | 9 - 10 | 1286 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppox
Genomic Map