Assay Details
| Gene Symbol: Sptlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009269.2 | NP_033295.2 | 5 - 6 | 439 | 62 | |
| GenBank mRNA | AF003823.1 | - | 5 - 6 | 443 | 62 | |
| AK028713.1 | - | 5 - 6 | 450 | 62 | ||
| AK053207.1 | - | 5 - 6 | 444 | 62 | ||
| AK079578.1 | - | 5 - 6 | 440 | 62 | ||
| AK084391.1 | - | 5 - 6 | 443 | 62 | ||
| AK084445.1 | - | 5 - 6 | 436 | 62 | ||
| AK089030.1 | - | 5 - 6 | 443 | 62 | ||
| AK161140.1 | - | 5 - 6 | 476 | 62 | ||
| BC046323.1 | - | 5 - 6 | 439 | 6391985 | 62 | |
| BC068243.1 | - | 3297 | 62 | |||
| X95641.1 | - | 5 - 6 | 438 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc1
Genomic Map