Assay Details
| Gene Symbol: Sptlc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009269.2 | NP_033295.2 | 11 - 12 | 1093 | 110 | |
| GenBank mRNA | AF003823.1 | - | 11 - 12 | 1097 | 110 | |
| AK028713.1 | - | 11 - 12 | 1104 | 110 | ||
| AK043064.1 | - | 7 - 8 | 672 | 110 | ||
| AK053207.1 | - | 11 - 12 | 1098 | 110 | ||
| AK079578.1 | - | 11 - 12 | 1094 | 110 | ||
| AK084391.1 | - | 11 - 12 | 1097 | 110 | ||
| AK084445.1 | - | 11 - 12 | 1090 | 110 | ||
| AK089030.1 | - | 11 - 12 | 1097 | 110 | ||
| AK161140.1 | - | 11 - 12 | 1130 | 110 | ||
| AK161811.1 | - | 7 - 8 | 672 | 110 | ||
| BC046323.1 | - | 11 - 12 | 1093 | 6391985 | 110 | |
| BC068243.1 | - | 3951 | 110 | |||
| X95641.1 | - | 11 - 12 | 1092 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sptlc1
Genomic Map