Assay Details
| Gene Symbol: Terf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009353.2 | NP_033379.2 | 7 - 8 | 1370 | 71 | |
| GenBank mRNA | AF003000.1 | - | 7 - 8 | 1264 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Terf2
Genomic Map