Assay Details
| Gene Symbol: Tnfaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159392.1 | NP_001152864.1 | 6 - 7 | 1008 | 121 | |
| NM_009395.4 | NP_033421.3 | 6 - 7 | 1021 | 121 | ||
| GenBank mRNA | AK004593.2 | - | 6 - 7 | 935 | 121 | |
| AK034305.1 | - | 6 - 7 | 956 | 121 | ||
| AK147809.1 | - | 6 - 7 | 916 | 121 | ||
| AK168386.1 | - | 6 - 7 | 900 | 121 | ||
| AK168704.1 | - | 6 - 7 | 943 | 121 | ||
| AK168805.1 | - | 6 - 7 | 917 | 121 | ||
| AK172287.1 | - | 3 - 4 | 264 | 121 | ||
| BC003906.1 | - | 6 - 7 | 857 | 3484017 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
cell migration
protein ubiquitination
negative regulation of Rho protein signal transduction
stress fiber assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA replication
protein homooligomerization
ubiquitin-protein transferase activity
GTP-Rho binding
protein domain specific binding
DNA replication
cell migration
protein ubiquitination
negative regulation of Rho protein signal transduction
stress fiber assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA replication
protein homooligomerization
ubiquitin-protein transferase activity
GTP-Rho binding
protein domain specific binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943939 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943937 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943938 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957845 | AK168805.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957845 | AK172287.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957845 | AK168704.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957845 | AK147809.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1957845 | AK034305.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86929 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696149 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 211137 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651074 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863264 | AK051498.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 346493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686418 | AK005667.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 318967 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 80897 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108767 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159186 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P173200 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957845 | AK168805.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957845 | AK172287.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957845 | AK168704.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957845 | AK147809.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1957845 | AK034305.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943925 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943916 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943926 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943919 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943915 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943924 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943938 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943921 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943917 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943932 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943923 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943930 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943928 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27943933 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07408582 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07408569 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159186 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P173200 | - | - |
| Whole Mouse Genome G4122A | A_52_P52036 | CB205355.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52036 | AK214142.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52036 | AK218103.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52036 | AK219473.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P52036 | AK168805.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131926 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159186 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P173200 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09218220 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09218201 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09218218 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09218209 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09218215 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943937 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943939 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943941 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943940 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P27943938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943847 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943900 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943803 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943876 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943869 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943917 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943923 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943907 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943827 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943899 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943916 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943897 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943820 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27943804 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09518822 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943915 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943932 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943907 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943903 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943926 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09218216 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943899 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943938 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943909 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09218202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09475094 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943923 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943934 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943898 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09083738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943919 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943912 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09475100 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943901 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943922 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943897 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943930 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943921 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09697628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943910 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943900 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943917 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27943940 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10388879 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388877 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10388872 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1448863_PM_a_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417866_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417865_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10388869 | - | - |
| Mouse Genome 430 2.0 Array | 1448863_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417865_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1448863_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1417865_at | - | - |
| Mouse Genome 430 2.0 Array | 1417866_at | - | - |
| Mouse Genome 430 2.0 Array | 1417866_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21927_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 21927_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100030430.mm.1 | NM_001159392.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030430.mm.1 | NM_001159392.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030430.mm.1 | NM_009395.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030430.mm.1 | NM_009395.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030430.mm.1 | TC1100003299.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100030431.mm.1 | NM_001159392.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030431.mm.1 | NM_001159392.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030431.mm.1 | NM_009395.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030431.mm.1 | NM_009395.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100030431.mm.1 | TC1100003299.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417865_PM_at | - | - |
| Mouse Expression Set 430 | 1448863_PM_a_at | - | - |
| Mouse Expression Set 430 | 1448863_a_at | - | - |
| Mouse Expression Set 430 | 1417865_at | - | - |
| Mouse Expression Set 430 | 1417866_at | - | - |
| Mouse Expression Set 430 | 1417866_PM_at | - | - |
| Murine Genome U74v2 Set | 93215_PM_at | - | - |
| Murine Genome U74v2 Set | 93215_at | - | - |
| Murine Genome U74v2 Set | 92572_at | - | - |
| Murine Genome U74v2 Set | 92572_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448863_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1448863_PM_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417866_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417865_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417866_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417865_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417866_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448863_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417865_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417865_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1448863_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417866_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6940593 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940593 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2429800 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2429800 | - | - |
| mouse-6 v1.1 (Illumina) | scl39881.7_182-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940593 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl39881.7_182-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940593 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2429800 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2429800 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl39881.7_182-S | - | - |

Tnfaip1
Genomic Map