Assay Details
| Gene Symbol: Ezr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009510.2 | NP_033536.2 | 1 - 2 | 152 | 65 | |
| GenBank mRNA | AK002766.1 | - | 1 - 2 | 126 | 65 | |
| AK004588.2 | - | 1 - 2 | 127 | 65 | ||
| AK135194.1 | - | 1 - 2 | 128 | 65 | ||
| AK135353.1 | - | 1 - 2 | 128 | 65 | ||
| AK159717.1 | - | 1 - 2 | 130 | 65 | ||
| AK159824.1 | - | 1 - 2 | 128 | 65 | ||
| AK159937.1 | - | 1 - 2 | 129 | 65 | ||
| AK168410.1 | - | 1 - 2 | 128 | 65 | ||
| AK168840.1 | - | 1 - 2 | 128 | 65 | ||
| AK170610.1 | - | 1 - 2 | 128 | 65 | ||
| AK172336.1 | - | 1 - 2 | 135 | 65 | ||
| BC048181.1 | - | 1 - 2 | 115 | 6826190 | 65 | |
| BC098502.1 | - | 1 - 2 | 104 | 30047137 | 65 | |
| X60671.1 | - | 1 - 2 | 110 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002766.1, AK135194.1, AK135353.1, AK159717.1, AK159824.1, AK168840.1, AK170610.1, AK172336.1, BC048181.1, BC098502.1, NM_009510.2, X60671.1 transcripts.
Set Membership: |
|

Ezr
Genomic Map