Assay Details
| Gene Symbol: Mpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010824.2 | NP_034954.2 | 1 - 2 | 291 | 60 | |
| GenBank mRNA | AY500847.1 | - | 1 - 2 | 45 | 60 | |
| CA547566.1 | - | 1 - 2 | 245 | 60 | ||
| X15313.1 | - | 1 - 2 | 501 | 60 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AY500847.1,NM_010824.2,X15313.1 transcripts.
Set Membership: |
|

Mpo
Genomic Map