Assay Details
| Gene Symbol: Ncf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286037.1 | NP_001272966.1 | 6 - 7 | 635 | 66 | |
| NM_010876.4 | NP_035006.3 | 6 - 7 | 635 | 66 | ||
| NR_104383.1 | 6 - 7 | 635 | 66 | |||
| GenBank mRNA | AB002663.1 | - | 6 - 7 | 614 | 66 | |
| AK149619.1 | - | 6 - 7 | 636 | 66 | ||
| AK149668.1 | - | 6 - 7 | 602 | 66 | ||
| AK149732.1 | - | 6 - 7 | 636 | 66 | ||
| AK150945.1 | - | 6 - 7 | 601 | 66 | ||
| AK151316.1 | - | 6 - 7 | 602 | 66 | ||
| AK151573.1 | - | 5 - 6 | 461 | 66 | ||
| AK151593.1 | - | 6 - 7 | 600 | 66 | ||
| AK152386.1 | - | 6 - 7 | 598 | 66 | ||
| AK162308.1 | - | 6 - 7 | 602 | 66 | ||
| AK162623.1 | - | 6 - 7 | 606 | 66 | ||
| AK170462.1 | - | 6 - 7 | 602 | 66 | ||
| AK171559.1 | - | 6 - 7 | 601 | 66 | ||
| BC026529.1 | - | 5 - 6 | 458 | 66 | ||
| L11455.1 | - | 6 - 7 | 572 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ncf1
Genomic Map