Assay Details
| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011074.2 | NP_035204.2 | 4 - 5 | 586 | 71 | |
| GenBank mRNA | AF033655.1 | - | 4 - 5 | 545 | 71 | |
| AK016370.1 | - | 4 - 5 | 528 | 71 | ||
| AK045083.1 | - | 4 - 5 | 587 | 71 | ||
| AK051283.1 | - | 4 - 5 | 560 | 71 | ||
| AK051411.1 | - | 4 - 5 | 529 | 71 | ||
| AK083269.1 | - | 3 - 4 | 741 | 71 | ||
| AK083790.1 | - | 4 - 5 | 529 | 71 | ||
| AK087398.1 | - | 3 - 4 | 357 | 71 | ||
| AK129226.1 | - | 4 - 5 | 413 | 71 | ||
| AK138437.1 | - | 3 - 4 | 827 | 71 | ||
| AK139386.1 | - | 3 - 4 | 747 | 71 | ||
| AK141535.1 | - | 3 - 4 | 515 | 71 | ||
| AK165149.1 | - | 3 - 4 | 431 | 71 | ||
| BC068134.1 | - | 3 - 4 | 790 | 6834018 | 71 | |
| U62391.1 | - | 3 - 4 | 440 | 71 | ||
More Information
Set Membership: |
|

Cdk14
Genomic Map