Assay Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284394.1 | NP_001271323.1 | 2 - 3 | 460 | 75 | |
| NM_011129.2 | NP_035259.1 | 2 - 3 | 460 | 75 | ||
| GenBank mRNA | AK039859.1 | - | 3 - 4 | 470 | 75 | |
| AK135143.1 | - | 2 - 3 | 461 | 75 | ||
| AK137299.1 | - | 2 - 3 | 379 | 75 | ||
| BC055101.1 | - | 2 - 3 | 460 | 6591016 | 75 | |
| BU562316.1 | - | 3 - 4 | 362 | 75 | ||
| X61452.1 | - | 2 - 3 | 390 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sept4
Genomic Map