Assay Details
| Gene Symbol: Sept4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284392.1 | NP_001271321.1 | 3 - 4 | 347 | 68 | |
| NM_001284394.1 | NP_001271323.1 | 4 - 5 | 644 | 68 | ||
| NM_001284398.1 | NP_001271327.1 | 3 - 4 | 331 | 68 | ||
| NM_011129.2 | NP_035259.1 | 4 - 5 | 644 | 68 | ||
| GenBank mRNA | AB078010.1 | - | 3 - 4 | 242 | 68 | |
| AK039859.1 | - | 5 - 6 | 654 | 68 | ||
| AK133933.1 | - | 3 - 4 | 299 | 68 | ||
| AK135143.1 | - | 4 - 5 | 645 | 68 | ||
| AK137299.1 | - | 4 - 5 | 563 | 68 | ||
| BC037673.1 | - | 3 - 4 | 349 | 68 | ||
| BC055101.1 | - | 4 - 5 | 644 | 6591016 | 68 | |
| BU562316.1 | - | 5 - 6 | 546 | 68 | ||
| X61452.1 | - | 4 - 5 | 574 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sept4
Genomic Map