Assay Details
| Gene Symbol: Prodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011172.2 | NP_035302.2 | 2 - 3 | 517 | 56 | |
| GenBank mRNA | AF120279.1 | - | 2 - 3 | 474 | 56 | |
| AK144068.1 | - | 2 - 3 | 487 | 56 | ||
| BC008548.1 | - | 2 - 3 | 341 | 56 | ||
| BC125327.1 | - | 2 - 3 | 320 | 40129842 | 56 | |
| U80020.1 | - | 2 - 3 | 466 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prodh
Genomic Map