Assay Details
| Gene Symbol: Prodh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011172.2 | NP_035302.2 | 8 - 9 | 1044 | 76 | |
| GenBank mRNA | AF120279.1 | - | 8 - 9 | 1001 | 76 | |
| AK144068.1 | - | 8 - 9 | 1014 | 76 | ||
| BC008548.1 | - | 8 - 9 | 868 | 76 | ||
| BC125327.1 | - | 8 - 9 | 847 | 40129842 | 76 | |
| U80020.1 | - | 8 - 9 | 993 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Prodh
Genomic Map