Assay Details
| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128605.1 | NP_001122077.1 | 3 - 4 | 438 | 68 | |
| NM_011183.3 | NP_035313.2 | 3 - 4 | 447 | 68 | ||
| GenBank mRNA | AF038935.1 | - | 2 - 3 | 375 | 68 | |
| AK014706.1 | - | 2 - 3 | 382 | 68 | ||
| AK148111.1 | - | 3 - 4 | 439 | 68 | ||
| AK154119.1 | - | 3 - 4 | 383 | 68 | ||
| BC010403.1 | - | 2 - 3 | 393 | 4038761 | 68 | |
| U57324.1 | - | 3 - 4 | 398 | 68 | ||
More Information
Set Membership: |
|

Psen2
Genomic Map