Assay Details
| Gene Symbol: Ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011201.3 | NP_035331.3 | 5 - 6 | 656 | 65 | |
| GenBank mRNA | AK149979.1 | - | 5 - 6 | 612 | 65 | |
| AK150322.1 | - | 5 - 6 | 612 | 65 | ||
| AK151371.1 | - | 5 - 6 | 569 | 65 | ||
| AK153088.1 | - | 5 - 6 | 594 | 65 | ||
| AK156281.1 | - | 5 - 6 | 617 | 65 | ||
| AK157431.1 | - | 5 - 6 | 657 | 65 | ||
| AK165567.1 | - | 5 - 6 | 621 | 65 | ||
| AK165949.1 | - | 6 - 7 | 736 | 65 | ||
| AK166020.1 | - | 5 - 6 | 612 | 65 | ||
| AK168255.1 | - | 5 - 6 | 594 | 65 | ||
| AK168772.1 | - | 5 - 6 | 611 | 65 | ||
| AK170620.1 | - | 5 - 6 | 612 | 65 | ||
| AK171380.1 | - | 5 - 6 | 612 | 65 | ||
| AK172206.1 | - | 6 - 7 | 729 | 65 | ||
| AK188339.1 | - | 2 - 3 | 157 | 65 | ||
| AK215902.1 | - | 2 - 3 | 156 | 65 | ||
| BC010191.1 | - | 5 - 6 | 574 | 3156854 | 65 | |
| M97590.1 | - | 5 - 6 | 572 | 65 | ||
| Z23057.1 | - | 2 - 3 | 186 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn1
Genomic Map