Assay Details
| Gene Symbol: Ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011201.3 | NP_035331.3 | 7 - 8 | 1030 | 61 | |
| GenBank mRNA | AK149979.1 | - | 7 - 8 | 986 | 61 | |
| AK150322.1 | - | 7 - 8 | 986 | 61 | ||
| AK151371.1 | - | 7 - 8 | 943 | 61 | ||
| AK153088.1 | - | 7 - 8 | 968 | 61 | ||
| AK156281.1 | - | 7 - 8 | 992 | 61 | ||
| AK157431.1 | - | 7 - 8 | 1031 | 61 | ||
| AK165567.1 | - | 7 - 8 | 995 | 61 | ||
| AK165949.1 | - | 8 - 9 | 1110 | 61 | ||
| AK166020.1 | - | 7 - 8 | 986 | 61 | ||
| AK168255.1 | - | 7 - 8 | 968 | 61 | ||
| AK168772.1 | - | 7 - 8 | 985 | 61 | ||
| AK170620.1 | - | 7 - 8 | 986 | 61 | ||
| AK171380.1 | - | 7 - 8 | 986 | 61 | ||
| AK172206.1 | - | 8 - 9 | 1103 | 61 | ||
| BC005729.1 | - | 2 - 3 | 383 | 61 | ||
| BC010191.1 | - | 7 - 8 | 948 | 3156854 | 61 | |
| M97590.1 | - | 7 - 8 | 946 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn1
Genomic Map