Assay Details
| Gene Symbol: Vipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011703.4 | NP_035833.2 | 11 - 12 | 1276 | 82 | |
| GenBank mRNA | AK052465.1 | - | 11 - 12 | 1277 | 82 | |
More Information
Set Membership: |
|

Vipr1
Genomic Map