Assay Details
| Gene Symbol: Slc27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011977.3 | NP_036107.1 | 4 - 5 | 770 | 88 | |
| GenBank mRNA | AK040923.1 | - | 3 - 4 | 709 | 88 | |
| AK153133.1 | - | 1 - 2 | 511 | 88 | ||
| AK155185.1 | - | 2 - 3 | 576 | 88 | ||
| AK164798.1 | - | 2 - 3 | 549 | 88 | ||
| AK165571.1 | - | 1 - 2 | 592 | 88 | ||
| BC028937.1 | - | 4 - 5 | 770 | 5362790 | 88 | |
| U15976.1 | - | 2 - 3 | 565 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a1
Genomic Map