Assay Details
| Gene Symbol: Slc27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011977.3 | NP_036107.1 | 6 - 7 | 1003 | 60 | |
| GenBank mRNA | AK040923.1 | - | 5 - 6 | 942 | 60 | |
| AK138548.1 | - | 3 - 4 | 974 | 60 | ||
| AK153133.1 | - | 3 - 4 | 744 | 60 | ||
| AK155185.1 | - | 4 - 5 | 809 | 60 | ||
| AK164798.1 | - | 4 - 5 | 782 | 60 | ||
| AK165571.1 | - | 3 - 4 | 825 | 60 | ||
| BC028937.1 | - | 6 - 7 | 1003 | 5362790 | 60 | |
| U15976.1 | - | 4 - 5 | 798 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a1
Genomic Map