Assay Details
| Gene Symbol: Slc27a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011977.3 | NP_036107.1 | 7 - 8 | 1093 | 73 | |
| GenBank mRNA | AK040923.1 | - | 6 - 7 | 1032 | 73 | |
| AK137700.1 | - | 1 - 2 | 190 | 73 | ||
| AK153133.1 | - | 4 - 5 | 834 | 73 | ||
| AK155185.1 | - | 5 - 6 | 899 | 73 | ||
| AK164798.1 | - | 5 - 6 | 872 | 73 | ||
| AK165571.1 | - | 4 - 5 | 915 | 73 | ||
| BC028937.1 | - | 7 - 8 | 1093 | 5362790 | 73 | |
| U15976.1 | - | 5 - 6 | 888 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a1
Genomic Map