Assay Details
| Gene Symbol: Slc27a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011978.2 | NP_036108.2 | 6 - 7 | 1392 | 77 | |
| GenBank mRNA | AF033031.1 | - | 6 - 7 | 1273 | 77 | |
| AF072757.1 | - | 6 - 7 | 1270 | 77 | ||
| AJ223958.1 | - | 6 - 7 | 1310 | 77 | ||
| AK144069.1 | - | 6 - 7 | 1394 | 77 | ||
| AK144070.1 | - | 6 - 7 | 1306 | 77 | ||
| AK165446.1 | - | 6 - 7 | 1393 | 77 | ||
| BC013442.1 | - | 6 - 7 | 1375 | 4236616 | 77 | |
| BC022170.1 | - | 6 - 7 | 1371 | 4159081 | 77 | |
| BC024735.1 | - | 6 - 7 | 1361 | 5123650 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a2
Genomic Map