Assay Details
| Gene Symbol: Slc27a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011978.2 | NP_036108.2 | 8 - 9 | 1685 | 84 | |
| GenBank mRNA | AF033031.1 | - | 8 - 9 | 1566 | 84 | |
| AF072757.1 | - | 8 - 9 | 1563 | 84 | ||
| AJ223958.1 | - | 8 - 9 | 1603 | 84 | ||
| AK144070.1 | - | 8 - 9 | 1599 | 84 | ||
| BC013442.1 | - | 8 - 9 | 1668 | 4236616 | 84 | |
| BC022170.1 | - | 8 - 9 | 1664 | 4159081 | 84 | |
| BC024735.1 | - | 8 - 9 | 1654 | 5123650 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc27a2
Genomic Map