| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100029067.mm.1 |
NM_001242580.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029067.mm.1 |
NM_001242580.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029067.mm.1 |
NM_012049.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029067.mm.1 |
NM_012049.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029067.mm.1 |
TC0100003541.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100029066.mm.1 |
NM_001242580.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029066.mm.1 |
NM_001242580.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029066.mm.1 |
NM_012049.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029066.mm.1 |
NM_012049.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029066.mm.1 |
TC0100003541.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100029075.mm.1 |
NM_001242580.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029075.mm.1 |
NM_001242580.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029075.mm.1 |
NM_012049.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029075.mm.1 |
NM_012049.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100029075.mm.1 |
TC0100003541.mm.1 |
MTA TC |