Assay Details
| Gene Symbol: Nit1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242580.1 | NP_001229509.1 | 6 - 7 | 819 | 100 | |
| NM_012049.2 | NP_036179.1 | 6 - 7 | 787 | 100 | ||
| GenBank mRNA | AF069988.1 | - | 6 - 7 | 768 | 100 | |
| AK004988.2 | - | 780 | 100 | |||
| AK032189.1 | - | 1470 | 100 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827416 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827415 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | BC021634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | AK004988.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | AK032189.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | NR_033728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | AF069988.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | BC021634.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | AK004988.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | AK032189.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | NR_033728.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P172532 | AF069988.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 393282 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110791 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | BC021634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | AK004988.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | AK032189.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | NR_033728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | AF069988.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | BC021634.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | AK004988.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | AK032189.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | NR_033728.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P172532 | AF069988.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | BC021634.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | AK004988.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | AK032189.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | NR_033728.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | AF069988.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | BC021634.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | AK004988.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | AK032189.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | NR_033728.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P172532 | AF069988.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827401 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827400 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827407 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827411 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827402 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827394 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827404 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827396 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P20827408 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110791 | - | - |
| Whole Mouse Genome G4122A | A_51_P172532 | BC021634.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | AK004988.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | AK032189.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | NR_033728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | AF069988.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | BC021634.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | AK004988.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | AK032189.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | NR_033728.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P172532 | AF069988.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322467 | AK004988.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322467 | AK032189.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322467 | BC023668.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322467 | BC054445.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P322467 | AK134547.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104177 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110791 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00516295 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00516296 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00516304 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P00516305 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827416 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P20827415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827390 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P20827404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00539774 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827414 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146821 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827413 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827404 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146825 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00516295 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P00539771 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827390 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827411 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827400 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P20827409 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01146820 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10360133 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360135 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360136 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360137 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360132 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10360134 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417469_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1417468_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10360130 | - | - |
| Mouse Genome 430 2.0 Array | 1417468_at | - | - |
| Mouse Genome 430 2.0 Array | 1417469_at | - | - |
| Mouse Genome 430 2.0 Array | 1417469_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1417468_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27045_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 27045_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0100029058.mm.1 | NM_001242580.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029058.mm.1 | NM_001242580.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029058.mm.1 | NM_012049.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029058.mm.1 | NM_012049.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0100029058.mm.1 | TC0100003541.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1417468_at | - | - |
| Mouse Expression Set 430 | 1417469_PM_at | - | - |
| Mouse Expression Set 430 | 1417469_at | - | - |
| Mouse Expression Set 430 | 1417468_PM_at | - | - |
| Murine Genome U74v2 Set | 160075_at | - | - |
| Murine Genome U74v2 Set | 160075_PM_at | - | - |
| Murine Genome U74v2 Set | 169303_at | - | - |
| Murine Genome U74v2 Set | 169303_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417468_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417469_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417469_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1417468_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417468_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417468_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417469_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1417469_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1230338 | - | - |
| MouseWG-6 v2 Expression BeadChip | 3940671 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3940671 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1230338 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003940671 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001230338 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2732145 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2754027 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2514451 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2732145 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2754027 | - | - |
| mouse-6 v1.1 (Illumina) | scl027045.1_0-S | - | - |
| mouse-6 v1.1 (Illumina) | scl000063.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1230338 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3940671 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl027045.1_0-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl000063.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1230338 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3940671 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2732145 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2754027 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2732145 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2754027 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl000063.1_0-S | - | - |

Nit1
Genomic Map