Assay Details
| Gene Symbol: Sgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001291152.1 | NP_001278081.1 | 2 - 3 | 307 | 67 | |
| NM_001291154.1 | NP_001278083.1 | 3 - 4 | 1023 | 67 | ||
| NM_013731.3 | NP_038759.1 | 3 - 4 | 422 | 67 | ||
| GenBank mRNA | AF169033.1 | - | 3 - 4 | 357 | 67 | |
| AK050009.1 | - | 3 - 4 | 1004 | 67 | ||
| AK136559.1 | - | 3 - 4 | 374 | 67 | ||
| BC026549.1 | - | 1 - 2 | 52 | 4973908 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sgk2
Genomic Map