Assay Details
| Gene Symbol: Park2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016694.3 | NP_057903.1 | 10 - 11 | 1262 | 115 | |
| GenBank mRNA | AB019558.1 | - | 10 - 11 | 1233 | 115 | |
| AF250293.1 | - | 10 - 11 | 1262 | 115 | ||
| BC113204.1 | - | 10 - 11 | 1245 | 40047891 | 115 | |
| FJ555554.1 | - | 8 - 9 | 1015 | 115 | ||
| FJ555555.1 | - | 5 - 6 | 673 | 115 | ||
| FJ555556.1 | - | 9 - 10 | 995 | 115 | ||
| FJ555557.1 | - | 9 - 10 | 1120 | 115 | ||
| GU272056.1 | - | 4 - 5 | 536 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Park2
Genomic Map