Assay Details
| Gene Symbol: Nudt5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016918.3 | NP_058614.1 | 6 - 7 | 773 | 66 | |
| GenBank mRNA | AF222786.1 | - | 5 - 6 | 423 | 66 | |
| AK088222.1 | - | 6 - 7 | 774 | 66 | ||
| BC004571.1 | - | 6 - 7 | 514 | 3586340 | 66 | |
| BC049595.1 | - | 6 - 7 | 578 | 6774692 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nudt5
Genomic Map