Assay Details
| Gene Symbol: Ppie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019489.5 | NP_062362.1 | 5 - 6 | 290 | 94 | |
| GenBank mRNA | AF182825.1 | - | 5 - 6 | 272 | 94 | |
| AK008177.1 | - | 5 - 6 | 291 | 94 | ||
| BC045154.1 | - | 5 - 6 | 285 | 6468589 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppie
Genomic Map