Assay Details
| Gene Symbol: Ppie | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019489.5 | NP_062362.1 | 9 - 10 | 844 | 61 | |
| GenBank mRNA | AF182825.1 | - | 9 - 10 | 826 | 61 | |
| AK008177.1 | - | 9 - 10 | 845 | 61 | ||
| AK042165.1 | - | 6 - 7 | 3428 | 61 | ||
| BC045154.1 | - | 9 - 10 | 839 | 6468589 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein peptidyl-prolyl isomerization
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
positive regulation of viral genome replication
nucleotide binding
nucleic acid binding
RNA binding
peptidyl-prolyl cis-trans isomerase activity
isomerase activity
poly(A) RNA binding
protein peptidyl-prolyl isomerization
regulation of transcription, DNA-templated
mRNA processing
protein folding
RNA splicing
positive regulation of viral genome replication
nucleotide binding
nucleic acid binding
RNA binding
peptidyl-prolyl cis-trans isomerase activity
isomerase activity
poly(A) RNA binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | AK042165.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | BC045154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | NM_019489.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | AF182825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | AK008177.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | AK042165.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | BC045154.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | NM_019489.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | AF182825.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P354433 | AK008177.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947985 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674035 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 358473 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 571854 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 185165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123223 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456195 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 181817 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 41462 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111686 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154319 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110789 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | AK042165.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | BC045154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | NM_019489.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | AF182825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | AK008177.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | AK042165.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | BC045154.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | NM_019489.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | AF182825.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P354433 | AK008177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P152132 | AK042165.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P152132 | BC045154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P152132 | NM_019489.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P152132 | AK008177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | AK042165.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | BC045154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | NM_019489.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | AF182825.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | AK008177.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | AK042165.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | BC045154.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | NM_019489.5 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | AF182825.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P354433 | AK008177.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261005 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261012 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23261015 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154319 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110789 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111686 | - | - |
| Whole Mouse Genome G4122A | A_52_P152132 | AK042165.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P152132 | BC045154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P152132 | NM_019489.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P152132 | AK008177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | AK042165.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | BC045154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | NM_019489.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | AF182825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | AK008177.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | AK042165.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | BC045154.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | NM_019489.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | AF182825.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P354433 | AK008177.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117089 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154319 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110789 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03534074 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03307834 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03534075 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03307832 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260984 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260945 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261020 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260928 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260887 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260883 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23261003 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04070197 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03658277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261005 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03609109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261019 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04070206 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03609105 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261031 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04070209 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03790267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261002 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03367211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03335589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03392930 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261023 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261010 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03989376 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260996 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04070199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03609101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260992 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03842515 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03849893 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03849895 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261003 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261034 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261006 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260999 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03849888 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260982 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03830664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261022 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261008 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261011 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04070211 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04038118 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03842512 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04038115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04048810 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260962 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23261015 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23260967 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10516082 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516085 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516087 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516083 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516088 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516090 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516086 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516089 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516091 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10516092 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450657_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10516081 | - | - |
| Mouse Genome 430 2.0 Array | 1450657_at | - | - |
| Mouse Genome 430 2.0 Array | 1450657_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56031_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 56031_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400028180.mm.1 | NM_019489.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028180.mm.1 | NM_019489.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028180.mm.1 | TC0400003478.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400028179.mm.1 | NM_019489.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028179.mm.1 | NM_019489.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028179.mm.1 | TC0400003478.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400028181.mm.1 | NM_019489.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028181.mm.1 | NM_019489.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400028181.mm.1 | TC0400003478.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450657_PM_at | - | - |
| Mouse Expression Set 430 | 1450657_at | - | - |
| Murine Genome U74v2 Set | 102278_at | - | - |
| Murine Genome U74v2 Set | 102278_PM_at | - | - |
| Murine Genome U74v2 Set | 168107_PM_i_at | - | - |
| Murine Genome U74v2 Set | 168107_i_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450657_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450657_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450657_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450657_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2260079 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260079 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1239400 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1239400 | - | - |
| mouse-6 v1.1 (Illumina) | scl23863.6.1_15-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260079 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl23863.6.1_15-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2970685 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2970691 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2970685 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2970691 | - | - |

Ppie
Genomic Map