Assay Details
| Gene Symbol: Trip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170907.1 | NP_001164378.1 | 3 - 4 | 547 | 73 | |
| NM_019797.4 | NP_062771.2 | 6 - 7 | 1363 | 73 | ||
| GenBank mRNA | AF197574.1 | - | 3 - 4 | 422 | 73 | |
| AF539614.1 | - | 6 - 7 | 1246 | 73 | ||
| AK039024.1 | - | 3 - 4 | 432 | 73 | ||
| AK078393.1 | - | 3 - 4 | 424 | 73 | ||
| AK146298.1 | - | 3 - 4 | 430 | 73 | ||
| AK150752.1 | - | 3 - 4 | 424 | 73 | ||
| BC021316.1 | - | 3 - 4 | 435 | 5027899 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trip4
Genomic Map