Assay Details
| Gene Symbol: Trip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170907.1 | NP_001164378.1 | 5 - 6 | 839 | 78 | |
| NM_019797.4 | NP_062771.2 | 8 - 9 | 1655 | 78 | ||
| GenBank mRNA | AF197574.1 | - | 5 - 6 | 714 | 78 | |
| AF539614.1 | - | 8 - 9 | 1538 | 78 | ||
| AK039024.1 | - | 5 - 6 | 724 | 78 | ||
| AK146298.1 | - | 5 - 6 | 722 | 78 | ||
| AK150752.1 | - | 5 - 6 | 716 | 78 | ||
| BC021316.1 | - | 5 - 6 | 727 | 5027899 | 78 | |
More Information
Set Membership: |
|

Trip4
Genomic Map