Assay Details
| Gene Symbol: Tdo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019911.2 | NP_064295.2 | 3 - 4 | 269 | 108 | |
| GenBank mRNA | AB476989.1 | - | 3 - 4 | 743 | 108 | |
| AB476990.1 | - | 3 - 4 | 629 | 108 | ||
| AK132271.1 | - | 3 - 4 | 292 | 108 | ||
| BC018390.1 | - | 3 - 4 | 269 | 4159877 | 108 | |
| U24493.1 | - | 3 - 4 | 256 | 108 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tdo2
Genomic Map