Assay Details
| Gene Symbol: Tdo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019911.2 | NP_064295.2 | 4 - 5 | 337 | 95 | |
| GenBank mRNA | AB476989.1 | - | 4 - 5 | 811 | 95 | |
| AB476990.1 | - | 4 - 5 | 697 | 95 | ||
| AK132271.1 | - | 4 - 5 | 360 | 95 | ||
| BC018390.1 | - | 4 - 5 | 337 | 4159877 | 95 | |
| U24493.1 | - | 4 - 5 | 324 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tdo2
Genomic Map