Assay Details
| Gene Symbol: Tdo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019911.2 | NP_064295.2 | 5 - 6 | 463 | 64 | |
| GenBank mRNA | AB476989.1 | - | 5 - 6 | 937 | 64 | |
| AB476990.1 | - | 5 - 6 | 823 | 64 | ||
| AK132271.1 | - | 5 - 6 | 486 | 64 | ||
| BC018390.1 | - | 5 - 6 | 463 | 4159877 | 64 | |
| S56366.1 | - | 1 - 2 | 121 | 64 | ||
| U24493.1 | - | 5 - 6 | 450 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tdo2
Genomic Map